Invitrogen
Manufacturer Code:PA542928
Catalog # PIPA542928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ST3GAL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 2,3-sialyltransferase ST3Gal II; Alpha 2,3-ST 2; alpha-2,3-sialyltransferase; beta-galactoside alpha-2,3-sialyltransferase 2; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; Monosialoganglioside sialyltransferase; Sialyltransferase 4B; sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4B (beta-galactoside alpha-2,3-sialyltransferase); sialyltransferase 5; sialyltransferase ST3Gal-II; SIAT4-B; ST3 beta-galactoside alpha-2 3-sialyltransferase 2 Gal-NAc6S SIAT4B ST3GALII ST3GalA.2; ST3Gal II; st3Gal II.2; st3gal2-r1; ST3GALA.2; ST3GalII
Gene Aliases: AI429591; AW822065; Gal-NAc6S; si:ch211-223o1.7; SIAT4B; Siat5; ST3GAL2; ST3GalA.2; ST3GALII
UniProt ID: (Human) Q16842, (Mouse) Q11204, (Rat) Q11205
Entrez Gene ID: (Human) 6483, (Bovine) 444879, (Dog) 489714, (Rabbit) 100339474, (Horse) 100054889, (Mouse) 20444, (Rat) 64442, (Guinea pig) 100721162, (Zebrafish) 445562
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.