Invitrogen
Manufacturer Code:PA5112275
Catalog # PIPA5112275
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived Human ST3GAL2 fragment |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-sialytransferase AI429591 alpha 23-sialyltransferase ST3Gal II Alpha 23-ST 2 alpha-23-sialyltransferase AW822065 beta-galactoside alpha-23-sialyltransferase 2 BOS_16927 CMP-N-acetylneuraminate-beta-galactosamide-alpha-23-sialyltransferase 2 Gal-beta-13-GalNAc-alpha-23-sialyltransferase Gal-NAc6S si:ch211-223o1.7 Sialyltransferase 4B sialyltransferase 4B (beta-galactosidase alpha-23-sialytransferase) sialyltransferase 4B (beta-galactoside alpha-23-sialyltransferase) sialyltransferase 5 sialyltransferase ST3Gal-II Siat4b SIAT4-B Siat5 ST3 beta-galactoside alpha-23-sialyltransferase 2 ST3Gal II st3Gal II.2 St3gal2 st3gal2-r1 ST3GalA.2 ST3GalII; Alpha 2,3-ST 2; beta-galactoside alpha-2,3-sialyltransferase 2; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; Monosialoganglioside sialyltransferase; Sialyltransferase 4B; sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase); SIAT4-B; ST3Gal II; ST3GalA.2; ST3GalII
Gene Aliases: Gal-NAc6S; SIAT4B; ST3GAL2; ST3GalA.2; ST3GALII
UniProt ID: (Human) Q16842
Entrez Gene ID: (Human) 6483
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.