Novus Biologicals
Manufacturer Code:NBP131449
Catalog # NBP131449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human SIAT4A. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha 2,3-ST 1; Alpha 23-ST 1 CMP-N-acetylneuraminate-beta-galactosamide-alpha-23-sialyltransferase 1 DKFZp666E036 DKFZp779K2051 EC 2.4.99 EC 2.4.99.4 Gal-beta-13-GalNAc-alpha-23-sialyltransferase Gal-NAc6S Sialyltransferase 4A sialyltransferase 4A (beta-galactosidase alpha-23-sialytransferase) sialyltransferase 4A (beta-galactoside alpha-23-sialyltransferase) sialyltransferase 4A (beta-galactoside alpha-23-sialytransferase) SIAT4 SIAT4A SIAT4-A SIATFLFLJ36548 ST3 beta-galactoside alpha-23-sialyltransferase 1 ST3Gal I ST3GalA.1MGC9183 ST3GalI ST3GalIA ST3GalIA1Beta-galactoside alpha-23-sialyltransferase 1 ST3OST3GalA; beta-galactoside alpha-2,3-sialyltransferase 1; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 1; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; Monosialoganglioside sialyltransferase; Sialyltransferase 4A; sialyltransferase 4A (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4A (beta-galactoside alpha-2,3-sialyltransferase); sialyltransferase 4A (beta-galactoside alpha-2,3-sialytransferase); SIAT4-A; SIATFL; ST3Gal I; ST3GalA.1; ST3GalI; ST3O
Gene Aliases: Gal-NAc6S; SIAT4; SIAT4A; SIATFL; ST3GAL1; ST3GalA; ST3GalA.1; ST3GalIA; ST3GalIA,1; ST3O
UniProt ID: (Human) Q11201
Entrez Gene ID: (Human) 6482
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.