R&D Systems
Manufacturer Code:MAB5232100
Catalog # MAB5232100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2154B |
Immunogen | <em>S. frugiperda</em> insect ovarian cell line<em> Sf</em> 21-derived recombinant human ST-2/ IL-33R Lys19-Phe328 Accession # Q01638 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: DER4 DER4ST2growth stimulation-expressed FIT-1 IL-1 R4 IL1R4 IL-1R4 IL-1RL1 IL-33 R IL33R IL-33R interleukin 1 receptor-like 1 interleukin 1 receptor-related protein interleukin-1 receptor-like 1 Ly84 MGC32623 Protein ST2 ST2L ST2V T1 T1homolog of mouse growth stimulation-expressed; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin-1 receptor-like 1; Protein ST2
Gene Aliases: DER4; FIT-1; IL1RL1; IL33R; ST2; ST2L; ST2V; T1
UniProt ID: (Human) Q01638
Entrez Gene ID: (Human) 9173
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.