Novus Biologicals
Manufacturer Code:NBP224736
Catalog # NBP224736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 25-65 of human ST2 protein was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: growth stimulation-expressed; homolog of mouse growth stimulation-expressed; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin-1 receptor-like 1; Protein ST2; suppression of tumorigenicity 2
Gene Aliases: DER4; FIT-1; IL1RL1; IL33R; ST2; ST2L; ST2V; T1
UniProt ID: (Human) Q01638
Entrez Gene ID: (Human) 9173
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.