Invitrogen
Manufacturer Code:PA523206
Catalog # PIPA523206
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (210 CKLKKSLIFSN 220) of ST2V protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DER4 FIT-1 IL33R ST2 ST2L ST2V T1 growth stimulation-expressed homolog of mouse growth stimulation-expressed interleukin 1 receptor-related protein interleukin-1 receptor-like 1; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Lymphocyte antigen 84; Protein ST2; Protein T1
Gene Aliases: DER4; FIT-1; IL1RL1; IL33R; Ly84; ST2; St2-rs1; ST2L; ST2V; Ste2; T1; T1/ST2
UniProt ID: (Human) Q01638, (Mouse) P14719
Entrez Gene ID: (Human) 9173, (Mouse) 17082
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.