Invitrogen
Manufacturer Code:PA5110727
Catalog # PIPA5110727
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human ST2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DER4 Fit1 Fit-1 Fos-responsive gene 1 protein IL-1 R4 Il1rl1 IL-1RL1 IL33R IL-33R Interleukin1 receptor like 1 Interleukin-1 receptor-like 1 Interleukin-33 receptor alpha chain Ly84 Lymphocyte antigen 84 Protein ST2 Protein T1 sIL 33R soluble IL 33 Receptor soluble IL 33R ST2 ST2L St2-rs1 ST2V Ste2 T1 T1/ST2; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin-1 receptor-like 1; Protein ST2
Gene Aliases: DER4; FIT-1; IL1RL1; IL33R; ST2; ST2L; ST2V; T1
UniProt ID: (Human) Q01638
Entrez Gene ID: (Human) 9173
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.