Invitrogen
Manufacturer Code:PA575323
Catalog # PIPA575323
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the N-terminual of Human ST14. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Epithin; Matriptase; matriptase/MT-SP1; membrane-type serine protease; Membrane-type serine protease 1; MT-SP1; Prostamin; protease, serine, 14 (epithin); Serine protease 14; Serine protease TADG-15; suppression of tumorigenicity 14 (colon carcinoma); suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin); Suppressor of tumorigenicity 14 protein; Suppressor of tumorigenicity 14 protein homolog; Suppressor of tumorigenicity 14 protein Matriptase Membrane-type serine protease 1 MT-SP1 Prostamin Serine protease 14 Serine protease TADG-15 Tumor-associated differentially-expressed gene 15 protein ST14 PRSS14 SNC19 TADG15; tumor associated differentially expressed gene 15 protein; Tumor-associated differentially-expressed gene 15 protein
Gene Aliases: ARCI11; Epithin; HAI; matriptase; mCAP3; MT-SP1; MTSP1; PRSS14; SNC19; ST14; TADG15; TMPRSS14
UniProt ID: (Human) Q9Y5Y6, (Mouse) P56677
Entrez Gene ID: (Human) 6768, (Rat) 114093, (Mouse) 19143
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.