Invitrogen
Manufacturer Code:PA547554
Catalog # PIPA547554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit human Matriptase/ST14 Catalytic Domain Val615-Val855 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: HAI MT-SP1 MTSP1 PRSS14 SNC19 TADG15 TMPRSS14 membrane-type serine protease 1 prostamin serine protease 14 serine protease TADG-15 suppression of tumorigenicity 14 (colon carcinoma matriptase epithin) suppressor of tumorigenicity 14 protein tumor associated differentially expressed gene 15 protein; Matriptase; Membrane-type serine protease 1; MT-SP1; Prostamin; Serine protease 14; Serine protease TADG-15; suppression of tumorigenicity 14 (colon carcinoma); suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin); Suppressor of tumorigenicity 14 protein; tumor associated differentially expressed gene 15 protein; Tumor-associated differentially-expressed gene 15 protein
Gene Aliases: ARCI11; HAI; MT-SP1; MTSP1; PRSS14; SNC19; ST14; TADG15; TMPRSS14
UniProt ID: (Human) Q9Y5Y6
Entrez Gene ID: (Human) 6768
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.