Novus Biologicals
Manufacturer Code:NBP274368UV
Catalog # NBP274368UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4D10 |
Immunogen | Full-length protein expressed in 293T cell transfected with human SSX2 expression vector. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Cancer/testis antigen 5.2; Cancer/testis antigen 5.2 cancer/testis antigen family 5 member 2a CT5.2a HD21 HOM-MEL-40 MGC119055 MGC15364 MGC3884 protein SSX2 sarcoma synovial X-chromosome-related 2 SSX SSX2A SSX2B synovial sarcoma X breakpoint 2B synovial sarcoma X breakpoint 2CT5.2 Tumor antigen HOM-MEL-40; cancer/testis antigen family 5, member 2a; CT5.2; Protein SSX2; sarcoma, synovial, X-chromosome-related 2; Synovial sarcoma, X breakpoint 2; synovial sarcoma, X breakpoint 2B; Tumor antigen HOM-MEL-40
Gene Aliases: CT5.2; CT5.2A; HD21; HOM-MEL-40; SSX; SSX2; SSX2A; SSX2B
UniProt ID: (Human) Q16385
Entrez Gene ID: (Human) 6757
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.