Invitrogen
Manufacturer Code:PA5112893
Catalog # PIPA5112893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Protein SSX2 protein (1-188AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 5.2; Cancer/testis antigen 5.2 cancer/testis antigen family 5 member 2a CT5.2 CT5.2A CT52 HD21 HOM-MEL-40 LOC103690782 MGC119055 MGC15364 MGC3884 Protein SSX2 putative protein SSX10 sarcoma synovial X-chromosome-related 2 SSX SSX family member 2 SSX2 SSX2A SSX2B Synovial sarcoma X breakpoint 2 synovial sarcoma X breakpoint 2B Tumor antigen HOM-MEL-40; cancer/testis antigen family 5, member 2a; CT5.2; Protein SSX2; sarcoma, synovial, X-chromosome-related 2; Synovial sarcoma, X breakpoint 2; synovial sarcoma, X breakpoint 2B; Tumor antigen HOM-MEL-40
Gene Aliases: CT5.2; CT5.2A; HD21; HOM-MEL-40; SSX; SSX2; SSX2A; SSX2B
UniProt ID: (Human) Q16385
Entrez Gene ID: (Human) 6757
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.