Invitrogen
Manufacturer Code:PA316736
Catalog # APSA6736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the C-terminus of rat SSR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C or -80°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: somatostatin receptor subtype 2; Somatostatin receptor type 2; somatotropin release-inhibiting factor receptor; SRIF-1; SS-2-R; SS2-R; SS2R
Gene Aliases: Smstr-2; Smstr2; Sst2; SSTR-2; SSTR2
UniProt ID: (Human) P30874, (Mouse) P30875, (Rat) P30680
Entrez Gene ID: (Human) 6752, (Mouse) 20606, (Rat) 54305
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.