Proteintech
Manufacturer Code:205871AP150UL
Catalog # 50-173-2976
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SSTR1 Fusion Protein Ag14287 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RNGPCRRNA; somatostatin receptor 1 Somatostatin receptor type 1 SRIF 2 SS 1 R SS1 R SS1R SSTR1; Somatostatin receptor subtype 1; Somatostatin receptor type 1; SRIF-2; SS-1-R; SS1-R; SS1R
Gene Aliases: Gpcrrna; Smstr-1; Smstr1; SRIF-2; SS-1-R; SS1-R; SS1R; sst1; SSTR1
UniProt ID: (Human) P30872, (Mouse) P30873, (Rat) P28646
Entrez Gene ID: (Human) 6751, (Mouse) 20605, (Rat) 25033
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.