R&D Systems
Manufacturer Code:MAB2155
Catalog # MAB2155
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MC-480 |
Immunogen | F9 mouse teratocarcinoma stem cells |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha (1,3) fucosyltransferase, myeloid-specific; Alpha-(1,3)-fucosyltransferase 4; alpha-(13)-fucosyltransferase CD15 EC 2.4.1 EC 2.4.1.- EC 2.4.1.65 ELAM-1 ligand fucosyltransferase ELFT FCT3AELAM ligand fucosyltransferase Fucosyltransferase 4 fucosyltransferase 4 (alpha (13) fucosyltransferase myeloid-specific) Fucosyltransferase IV fucT-IV FUC-TIV FUTIV Galactoside 3-L-fucosyltransferase Lewis X LeX SSEA1 SSEA-1 stage-specific embryonic antigen 1; ELAM ligand fucosyltransferase; ELAM-1 ligand fucosyltransferase; Fuc-TIV; Fucosyltransferase 4; fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific); Fucosyltransferase IV; fucT-IV; Galactoside 3-L-fucosyltransferase; Lewis X; stage-specific embryonic antigen 1
Gene Aliases: CD15; ELFT; FCT3A; FUC-TIV; FUT4; FUTIV; LeX; SSEA-1
UniProt ID: (Human) P22083
Entrez Gene ID: (Human) 2526
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.