OriGene
Manufacturer Code:TA500426
Catalog # 50-167-1561
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C8 |
Immunogen | Full-length protein expressed in 293T cell transfected with human SSB expression vector |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: autoantigen La; La autoantigen; La ribonucleoprotein; La ribonucleoprotein domain family, member 3; LARP3 La La/SSB; lupus La antigen; Lupus La protein; Sjoegren syndrome type B antigen; Sjogren syndrome antigen B; Sjogren syndrome antigen B (autoantigen La); SS-B; SS-B/La protein
Gene Aliases: La; La/SSB; LARP3; SSB
UniProt ID: (Human) P05455
Entrez Gene ID: (Dog) 478787, (Human) 6741
Molecular Function: RNA binding protein nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.