Invitrogen
Manufacturer Code:PA575246
Catalog # PIPA575246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 50-100 of Human SRY. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: essential protein for sex determination in human males; HMG box transcription factor Sry1; HMG box transcription factor Sry3BI; sex determining region of Chr Y; sex determining region on Y; sex determining region Y protein; sex-determining region on Y; Sex-determining region Y protein; Sex-determining region Y protein Testis-determining factor SRY TDF; Testis determining locus; testis determining-Y; Testis-determining factor; testis-determining factor on Y
Gene Aliases: SRXX1; SRXY1; SRY; Sry1; Sry3BI; SRYGENE; TDF; TDY
UniProt ID: (Human) Q05066, (Mouse) Q05738, (Rat) P36394
Entrez Gene ID: (Human) 6736, (Mouse) 21674, (Rat) 25221
Molecular Function: HMG box transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.