Invitrogen
Manufacturer Code:PA5113599
Catalog # PIPA5113599
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SRY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: essential protein for sex determination in human males; essential protein for sex determination in human males HMG box transcription factor Sry1 HMG box transcription factor Sry3BI sex determining region of Chr Y sex determining region on Y sex determining region Y sex determining region Y protein sex-determining region on Y Sex-determining region Y protein SRXX1 SRXY1 SRY Sry1 Sry3BI SRYGENE TDF Tdy Testis determining locus testis determining-Y Testis-determining factor testis-determining factor on Y; sex determining region Y protein; sex-determining region on Y; Sex-determining region Y protein; Testis-determining factor; testis-determining factor on Y
Gene Aliases: SRXX1; SRXY1; SRY; TDF; TDY
UniProt ID: (Human) Q05066
Entrez Gene ID: (Human) 6736
Molecular Function: HMG box transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.