Invitrogen
Manufacturer Code:PA544514
Catalog # PIPA544514
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SFRS7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aging-associated protein 3; Serine/arginine-rich splicing factor 7; serine/arginine-rich splicing factor 7-like protein; Splicing factor 9G8; Splicing factor, arginine/serine-rich 7; splicing factor, arginine/serine-rich 7, 35kDa; SR splicing factor 7
Gene Aliases: 35kDa; 9430065L19Rik; 9G8; AAG3; NX-96; SFRS7; SRSF7
UniProt ID: (Human) Q16629, (Bovine) Q3T106, (Mouse) Q8BL97
Entrez Gene ID: (Horse) 100054033, (Human) 6432, (Bovine) 507066, (Dog) 475730, (Rabbit) 103347700, (Mouse) 225027, (Guinea pig) 100714040, (Rat) 362687
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.