Invitrogen
Manufacturer Code:PA562282
Catalog # PIPA562282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SRSF4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: arginine/serine-rich 4 Pre mRNA splicing factor SRP75 Pre-mRNA-splicing factor SRP75 Serine/arginine-rich splicing factor 4 Splicing factor Splicing factor arginine/serine rich 4 SRP001LB SRP75 Srsf4; Pre-mRNA-splicing factor SRP75; Serine/arginine-rich splicing factor 4; Splicing factor, arginine/serine-rich 4; SR splicing factor 4; SRP001LB
Gene Aliases: SFRS4; SRP75; SRSF4
UniProt ID: (Human) Q08170
Entrez Gene ID: (Human) 6429
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.