Invitrogen
Manufacturer Code:MA534705
Catalog # PIMA534705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JB40-76 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 1110054N12Rik 5730507C05Rik 6330415C05Rik AI482334 Alternative-splicing factor 1 ASF ASF/SF2 ASF-1 AW491331 OK/SW-cl.3 pre-mRNA-splicing factor SF2 P33 subunit Pre-mRNA-splicing factor SRp30a Serine/arginine-rich splicing factor 1 SF2 SF2P33 SFRS1 Splicing factor arginine/serine-rich 1 SRp30a Srsf1; Alternative-splicing factor 1; ASF-1; pre-mRNA-splicing factor SF2, P33 subunit; Serine/arginine-rich splicing factor 1; splicing factor 2; Splicing factor, arginine/serine-rich 1; splicing factor, arginine/serine-rich, 30-KD, A; SR splicing factor 1
Gene Aliases: ASF; OK/SW-cl.3; SF2; SF2P33; SFRS1; SRp30a; SRSF1
UniProt ID: (Human) Q07955
Entrez Gene ID: (Human) 6426
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.