Invitrogen
Manufacturer Code:PA530220
Catalog # PIPA530220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 224 of Human SF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alternative-splicing factor 1; ASF SF2 SF2p33 SFRS1 SRp30a ASF-1 SR splicing factor 1 alternative-splicing factor 1 pre-mRNA-splicing factor SF2 P33 subunit splicing factor 2 splicing factor arginine/serine-rich 1; ASF-1; ASF/SF2; pre-mRNA-splicing factor SF2, P33 subunit; Pre-mRNA-splicing factor SRp30a; Serine/arginine-rich splicing factor 1; splicing factor 2; Splicing factor, arginine/serine-rich 1; splicing factor, arginine/serine-rich 1 (ASF/SF2); splicing factor, arginine/serine-rich, 30-KD, A; SR splicing factor 1
Gene Aliases: 1110054N12Rik; 5730507C05Rik; 6330415C05Rik; AI482334; ASF; AW491331; OK/SW-cl.3; SF2; SF2P33; SFRS1; SRp30a; SRSF1
UniProt ID: (Human) Q07955, (Mouse) Q6PDM2
Entrez Gene ID: (Human) 6426, (Mouse) 110809
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.