Abnova
Manufacturer Code:H00063826D01
Catalog # 89-002-926
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SRR (NP_068766.1 1 a.a. ~ 340 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human SRR protein.
For Research Use Only
Protein Aliases: D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase
Gene Aliases: ILV1; ISO1; SRR
UniProt ID: (Human) Q9GZT4
Entrez Gene ID: (Human) 63826
Molecular Function:
deaminase
dehydratase
epimerase/racemase
hydrolase
isomerase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.