Novus Biologicals
Manufacturer Code:NBP29477420UL
Catalog # NB024065
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 166-465 of human SRPX2 (NP_055282.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BPP CBPS RESDX SRPULPMGX sushi repeat-containing protein SRPX2 sushi-repeat containing protein X-linked 2 sushi-repeat protein up-regulated in leukemia sushi-repeat-containing protein X-linked 2; Sushi repeat-containing protein SRPX2; sushi-repeat containing protein, X-linked 2; sushi-repeat protein up-regulated in leukemia; Sushi-repeat protein upregulated in leukemia; sushi-repeat-containing protein, X-linked 2
Gene Aliases: BPP; CBPS; PMGX; RESDX; SRPUL; SRPX2
UniProt ID: (Human) O60687
Entrez Gene ID: (Human) 27286
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.