Invitrogen
Manufacturer Code:PA5116545
Catalog # PIPA5116545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 40-320 of human SRPX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Down-regulated by v-SRC Drs DRS protein drs-1 drs-2 epididymis secretory sperm binding protein Li 83p ETX1 HEL-S-83p mDRS-1 mDRS-2 SRPX SRPX1 sushi repeat containing protein X-linked sushi repeat-containing protein SRPX sushi-repeat containing protein X-linked sushi-repeat-containing protein sushi-repeat-containing protein SRPX sushi-repeat-containing protein X chromosome sushi-repeat-containing protein X chromosome sushi-repeat-containing protein X-chromosome sushi-repeat-containing protein X-linked; DRS protein; epididymis secretory sperm binding protein Li 83p; Sushi repeat-containing protein SRPX; sushi-repeat containing protein, X-linked; Sushi-repeat-containing protein SRPX; sushi-repeat-containing protein, X chromosome; sushi-repeat-containing protein, X-chromosome; sushi-repeat-containing protein, X-linked
Gene Aliases: DRS; drs-1; drs-2; ETX1; HEL-S-83p; mDRS-1; mDRS-2; SRPX; SRPX1
UniProt ID: (Human) P78539, (Mouse) Q9R0M3
Entrez Gene ID: (Human) 8406, (Mouse) 51795
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.