Novus Biologicals
Manufacturer Code:NBP286829
Catalog # NB034451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Mouse SRPX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DRS ETX1SRPX1 sushi repeat-containing protein SRPX sushi-repeat containing protein X-linked sushi-repeat-containing protein X chromosome sushi-repeat-containing protein X-linked; epididymis secretory sperm binding protein Li 83p; Sushi repeat-containing protein SRPX; sushi-repeat containing protein, X-linked; sushi-repeat-containing protein, X chromosome; sushi-repeat-containing protein, X-linked
Gene Aliases: DRS; ETX1; HEL-S-83p; SRPX; SRPX1
UniProt ID: (Human) P78539
Entrez Gene ID: (Human) 8406
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.