Abnova
Manufacturer Code:PAB16858
Catalog # 89-118-672
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus 13 amino acids of human SRPX. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SRPX.
For Research Use Only
Protein Aliases: DRS; epididymis secretory sperm binding protein Li 83p; ETX1; SRPX1; Sushi repeat-containing protein SRPX; sushi-repeat containing protein, X-linked; sushi-repeat-containing protein, X chromosome; sushi-repeat-containing protein, X-linked
Gene Aliases: DRS; ETX1; HEL-S-83p; SRPX; SRPX1
UniProt ID: (Human) P78539
Entrez Gene ID: (Human) 8406
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.