Novus Biologicals
Manufacturer Code:NBP29362620UL
Catalog # NB029117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human SRPR (NP_003130.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Docking protein alpha; Docking protein alpha DP DP-alpha MGC17355 MGC3650 MGC9571 signal recognition particle receptor (docking protein) signal recognition particle receptor (&primedocking protein&prime) signal recognition particle receptor subunit alpha Sralpha SR-alpha SRP-alpha; DP-alpha; signal recognition particle receptor (docking protein); Signal recognition particle receptor subunit alpha; SR-alpha; SRP-alpha
Gene Aliases: DP; Sralpha; SRPR; SRPRA
UniProt ID: (Human) P08240
Entrez Gene ID: (Human) 6734
Molecular Function: G-protein RNA binding protein enzyme modulator nucleic acid binding receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.