Proteintech
Manufacturer Code:660681IG150UL
Catalog # 50-173-6505
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H7G10 |
Immunogen | SRP9 Fusion Protein Ag17153 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ALURBP SRP9; Signal recognition particle 9 kDa protein; signal recognition particle 9kD; signal recognition particle 9kDa; SRP9
Gene Aliases: 9kDa; ALURBP; SRP9
UniProt ID: (Human) P49458, (Mouse) Q9D085
Entrez Gene ID: (Human) 6726, (Rat) 690345, (Mouse) 27058
Molecular Function:
RNA binding protein
nucleic acid binding
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.