Novus Biologicals
Manufacturer Code:NBP261107
Catalog # NBP261107
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A region between residue 36 to 86 of human Signal recognition particle 9 kDa protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: ALURBP MGC110981 signal recognition particle 9 kDa protein signal recognition particle 9kD signal recognition particle 9kDa; Signal recognition particle 9 kDa protein; signal recognition particle 9kD; signal recognition particle 9kDa; SRP9
Gene Aliases: ALURBP; SRP9
UniProt ID: (Human) P49458
Entrez Gene ID: (Human) 6726
Molecular Function:
RNA binding protein
nucleic acid binding
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.