Invitrogen
Manufacturer Code:PA556994
Catalog # PIPA556994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SRP72 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis luminal protein 103; Signal recognition particle 72 kDa protein; signal recognition particle 72kD; signal recognition particle 72kDa; Signal recognition particle subunit SRP72; Signal recognition particle subunit SRP72 SRP72 Signal recognition particle 72 kDa protein SRP72; SRP72
Gene Aliases: BMFF; BMFS1; HEL103; SRP72
UniProt ID: (Human) O76094
Entrez Gene ID: (Human) 6731
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.