Invitrogen
Manufacturer Code:PA541843
Catalog # PIPA541843
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SRP68 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 68kDA subunit of signal recognition particle; likely ortholog of H. sapiens signal recognition particle 68kDa (SRP68); Signal recognition particle 68 kDa protein; signal recognition particle 68kD; Signal recognition particle 68kDa; Signal recognition particle subunit SRP68; SRP68
Gene Aliases: 2610024I03Rik; SRP68; zgc:92573
UniProt ID: (Dog) Q00004, (Human) Q9UHB9, (Mouse) Q8BMA6
Entrez Gene ID: (Pig) 100739290, (Bovine) 538777, (Horse) 100051325, (Goat) 102188706, (Dog) 403952, (Human) 6730, (Rabbit) 100356249, (Rat) 363707, (Mouse) 217337, (Guinea pig) 100721562, (Zebrafish) 446119
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.