Invitrogen
Manufacturer Code:PA5104335
Catalog # PIPA5104335
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SRP54 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 54K subunit of signal recognition particle; 54K subunit of signal recognition particle 54kDa signal recognition particle 54 signal recognition particle 54 kDa protein signal recognition particle 54A signal recognition particle 54kD signal recognition particle 54kDa Srp54 Srp54a; Signal recognition particle 54 kDa protein; signal recognition particle 54kD; signal recognition particle 54kDa; SRP54
Gene Aliases: 54kDa; SRP54
UniProt ID: (Human) P61011, (Mouse) P14576, (Rat) Q6AYB5
Entrez Gene ID: (Human) 6729, (Mouse) 24067, (Rat) 116650
Molecular Function:
G-protein
RNA binding protein
enzyme modulator
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.