Proteintech
Manufacturer Code:117291AP150UL
Catalog # 50-172-7408
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SRP54 Fusion Protein Ag2327 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 54K subunit of signal recognition particle; Signal recognition particle 54 kDa protein; signal recognition particle 54kD; signal recognition particle 54kDa; SRP54
Gene Aliases: 54kDa; SRP54
UniProt ID: (Human) P61011, (Mouse) P14576, (Rat) Q6AYB5
Entrez Gene ID: (Human) 6729, (Mouse) 24067, (Rat) 116650
Molecular Function:
G-protein
RNA binding protein
enzyme modulator
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.