Invitrogen
Manufacturer Code:PA541768
Catalog # PIPA541768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SRP14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (homologous Alu RNA binding protein); 18 kDa Alu RNA-binding protein; homologous Alu RNA binding protein; homologous Alu RNA-binding protein; Signal recognition particle 14 kDa protein; signal recognition particle 14kDa (homologous Alu RNA binding protein); Signal recognition particle 14kDa (homologous Alu RNA binding protein) ALURBP; SRP14
Gene Aliases: 14kDa; ALURBP; AW536328; SRP14; zgc:112290
UniProt ID: (Dog) P16255, (Human) P37108, (Bovine) A6QQL9, (Mouse) P16254
Entrez Gene ID: (Dog) 403930, (Horse) 100057403, (Human) 6727, (Rabbit) 100358027, (Bovine) 512792, (Sheep) 100135456, (Mouse) 20813, (Rat) 296076, (Guinea pig) 100713306, (Zebrafish) 619260
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.