Proteintech
Manufacturer Code:164711AP150UL
Catalog # 50-173-0967
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SRI Fusion Protein Ag9553 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 22 kDa protein; 22 kDa protein CP 22 SCN sorcin SRI V19; calcium binding protein amplified in mutlidrug-resistant cells; CP-22; CP22; H_RG167B05.1; Sorcin; V19
Gene Aliases: 2210417O06Rik; 2900070H08Rik; CP-22; CP22; SCN; Sor; SRI; V19
UniProt ID: (Human) P30626, (Mouse) Q6P069
Entrez Gene ID: (Human) 6717, (Mouse) 109552, (Rat) 683667
Molecular Function:
annexin
calcium-binding protein
calmodulin
cysteine protease
hydrolase
intracellular calcium-sensing protein
protease
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.