Novus Biologicals
Manufacturer Code:NBP300168UV
Catalog # NB064716
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant SRGN (CAA34900.1 Met1-Leu158). |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ12930 Hematopoetic proteoglycan core protein MGC9289 P.PG Platelet proteoglycan core protein platelet proteoglycan protein core PPG PRG1p.PG PRGhematopoetic proteoglycan core peptide proteoglycan 1 secretory granule proteoglycan protein core for mast cell secretory granule secretory granule proteoglycan 1 secretory granule proteoglycan core peptide Secretory granule proteoglycan core protein serglycin serglycin proteoglycan; hematopoetic proteoglycan core peptide; hematopoetic proteoglycan core protein; Hematopoietic proteoglycan core protein; P.PG; Platelet proteoglycan core protein; proteoglycan 1, secretory granule; proteoglycan protein core for mast cell secretory granule; secretory granule proteoglycan core peptide; Secretory granule proteoglycan core protein; Serglycin; serglycin proteoglycan
Gene Aliases: PPG; PRG; PRG1; SRGN
UniProt ID: (Human) P10124
Entrez Gene ID: (Human) 5552
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.