Novus Biologicals
Manufacturer Code:NBP188830
Catalog # NBP188830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:WEGRHNGIDGLIPHQYIVVQDTEDGVVERSSPKSEIEVISEPPEEKVTARAGASCPSGGHVADIYLANINKQRKRPESGSIR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FNBP2 Formin-binding protein 2 KIAA0456ARHGAP34formin binding protein 2 Rho GTPase-activating protein 34 SLIT-ROBO Rho GTPase activating protein 2 SLIT-ROBO Rho GTPase-activating protein 2 srGAP2 SRGAP3; Formin-binding protein 2; Rho GTPase-activating protein 34; SLIT-ROBO GAP2; SLIT-ROBO Rho GTPase-activating protein 2; srGAP2
Gene Aliases: ARHGAP34; FNBP2; KIAA0456; SRGAP2; SRGAP2A; SRGAP3
UniProt ID: (Human) O75044
Entrez Gene ID: (Human) 23380
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.