Proteintech
Manufacturer Code:132521AP150UL
Catalog # 50-172-8603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SRGAP1 Fusion Protein Ag4104 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARHGAP13 KIAA1304 SRGAP1; Rho GTPase activating protein 13; Rho GTPase-activating protein 13; SLIT-ROBO Rho GTPase-activating protein 1; srGAP1
Gene Aliases: 4930572H05Rik; ARHGAP13; KIAA1304; NMTC2; RGD1566260; SRGAP1
UniProt ID: (Human) Q7Z6B7, (Mouse) Q91Z69
Entrez Gene ID: (Human) 57522, (Rat) 314903, (Mouse) 117600
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.