R&D Systems
Manufacturer Code:MAB7119SP
Catalog # MAB7119SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 751512 |
Immunogen | <EM>E. coli</EM>-derived recombinant human SREBP2 Leu242-Asp450 Accession # Q12772 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHd2; BHLHD2 bHLHd2sterol regulatory element-binding protein 2 Class D basic helix-loop-helix protein 2 SREBF2 SREBP2 SREBP2SREBP-2 sterol regulatory element binding transcription factor 2 Sterol regulatory element-binding transcription factor 2; Class D basic helix-loop-helix protein 2; Processed sterol regulatory element-binding protein 2; SREBP-2; Sterol regulatory element-binding protein 2; Sterol regulatory element-binding transcription factor 2; Transcription factor SREBF2
Gene Aliases: BHLHD2; SREBF2; SREBP-2; SREBP2
UniProt ID: (Human) Q12772
Entrez Gene ID: (Human) 6721
Molecular Function:
basic helix-loop-helix transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.