Invitrogen
Manufacturer Code:PA588943
Catalog # PIPA588943
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-220 of human SREBF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHd2; Class D basic helix-loop-helix protein 2; Processed sterol regulatory element-binding protein 2; srebf2; srebf2 {ECO:0000312|EMBL:AAI01903.1}; SREBP-2; sterol regulatory element binding factor 2; sterol regulatory element binding protein 2; Sterol regulatory element-binding protein 2; Sterol regulatory element-binding transcription factor 2; Transcription factor SREBF2
Gene Aliases: AI608257; BHLHD2; lop13; nuc; SREBF2; Srebf2_retired; SREBP-2; SREBP2; SREBP2gc
UniProt ID: (Human) Q12772, (Rat) Q3T1I5, (Mouse) Q3U1N2
Entrez Gene ID: (Human) 6721, (Rat) 300095, (Mouse) 20788
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.