Invitrogen
Manufacturer Code:PA572661
Catalog # PIPA572661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 441-473 amino acids from the Central region of human (DANRE) srebf2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Processed sterol regulatory element-binding protein 2; SREBP-2; Sterol regulatory element-binding protein 2; Sterol regulatory element-binding protein 2 SREBP-2 Sterol regulatory element-binding transcription factor 2 Processed sterol regulatory element-binding protein 2 srebf2 {ECO:0000250; Sterol regulatory element-binding transcription factor 2; Transcription factor SREBF2; UniProtKB:Q12772}
Gene Aliases: srebf2; SREBP2; zgc:158371
UniProt ID: (Zebrafish) A3KNA7
Entrez Gene ID: (Zebrafish) 100037309
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.