Invitrogen
Manufacturer Code:MA516124
Catalog # PIMA516124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A4 |
Immunogen | 6 His-tag fusion protein of SREBP1 corresponding to amino acids 301-407 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ADD1; Adipocyte determination- and differentiation-dependent factor 1; bHLHd1; Class D basic helix-loop-helix protein 1; Processed sterol regulatory element-binding protein 1; SREBP-1; SREBP-1c SREBP1 bHLHd1 SREBP-1 class D basic helix-loop-helix protein 1 sterol regulatory element-binding protein 1 nt-SREBP1; SREBP1; sterol regulatory element binding protein 1; sterol regulatory element binding protein-1; sterol regulatory element binding-protein 1; Sterol regulatory element-binding protein 1; Sterol regulatory element-binding transcription factor 1; Transcription factor SREBF1
Gene Aliases: ADD-1; ADD1; BHLHD1; D630008H06; I79_010155; SREBF1; SREBP-1; SREBP-1a; SREBP-1c; SREBP1; SREBP1a; SREBP1c
UniProt ID: (Human) P36956, (Rat) P56720, (Chinese hamster) Q60416, (Mouse) Q9WTN3
Entrez Gene ID: (Human) 6720, (Chicken) 373915, (Dog) 403475, (Rat) 78968, (Chinese hamster) 100689018, (Mouse) 20787
Molecular Function:
basic helix-loop-helix transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.