Invitrogen
Manufacturer Code:MA511685
Catalog # PIMA511685
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A4 |
Immunogen | Recombit protein encoding N-terminal amino acids 301-407 (the bHLH/leucine zipper domain) of human SREBP-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHd1; Class D basic helix-loop-helix protein 1; Processed sterol regulatory element-binding protein 1; SREBP-1; SREBP-1 class D basic helix-loop-helix protein 1 sterol regulatory element binding protein-1 sterol regulatory element-binding protein 1 sterol regulatory element-binding transcription factor 1 nt-SREBP1; sterol regulatory element binding protein-1; Sterol regulatory element-binding protein 1; Sterol regulatory element-binding transcription factor 1; Transcription factor SREBF1
Gene Aliases: BHLHD1; I79_010155; SREBF1; SREBP-1; SREBP-1c; SREBP1; SREBP1a
UniProt ID: (Human) P36956, (Chinese hamster) Q60416
Entrez Gene ID: (Human) 6720, (Chinese hamster) 100689018
Molecular Function:
basic helix-loop-helix transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.