Invitrogen
Manufacturer Code:PA5107029
Catalog # PIPA5107029
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide located within the following region: GLLPGCAIFQ DLIRYGKTKC GEPSRPAACR AFDVPKRYFS HFYIISVLWN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-oxo-5-alpha-steroid 4-dehydrogenase 3; Polyprenol reductase; probable polyprenol reductase; S5AR 3; SR type 3; Steroid 5-alpha-reductase 2-like; Steroid 5-alpha-reductase 3
Gene Aliases: CDG1P; CDG1Q; KRIZI; SRD5A2L; SRD5A2L1; SRD5A3
UniProt ID: (Human) Q9H8P0
Entrez Gene ID: (Human) 79644
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.