Invitrogen
Manufacturer Code:MA537985
Catalog # PIMA537985
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2287 |
Immunogen | A synthesized peptide derived from human SRD5A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 3-oxo-5-alpha-steroid 4-dehydrogenase 2; 3-oxo-5-alpha-steroid 4-dehydrogenase 2 5 alpha-SR2 5art2 MGC138457 S5AR 2 SR type 2 Srd5a2 steroid 5 alpha reductase type2 steroid 5 alpha-reductase 2 steroid 5-alpha-reductase 2 steroid-5-alpha-reductase alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2) type II 5-alpha reductase; 5 alpha-SR2; S5AR 2; SR type 2; steroid 5 alpha reductase type2; Steroid 5-alpha-reductase 2; steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2); Type II 5-alpha reductase
Gene Aliases: 5ART2; S5AR 2; SRD5A2
UniProt ID: (Human) P31213, (Rat) P31214, (Mouse) Q99N99
Entrez Gene ID: (Human) 6716, (Rat) 64677, (Mouse) 94224
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.