Invitrogen
Manufacturer Code:PA575919
Catalog # PIPA575919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human SRD5A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-oxo-5-alpha-steroid 4-dehydrogenase 1; 3-oxo-5-alpha-steroid 4-dehydrogenase 1 (EC:1.3.1.22) SR type 1 Steroid 5-alpha-reductase 1 S5AR 1 SRD5A1; S5AR 1; SR type 1; steroid 5 alpha-reductase 1; Steroid 5-alpha-reductase 1; steroid 5-alpha-reductase type I; steroid-5-alpha-reductase 1; steroid-5-alpha-reductase, alpha polypeptide 1 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 1); type 1 5alpha-reductase
Gene Aliases: 0610031P22Rik; 4930435F02Rik; S5AR 1; Srd5a-1; SRD5A1
UniProt ID: (Human) P18405, (Mouse) Q505K7, (Rat) P24008
Entrez Gene ID: (Human) 6715, (Mouse) 78925, (Rat) 24950
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.