Novus Biologicals
Manufacturer Code:NBP305266
Catalog # NB094659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human SRD5A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-oxo-5-alpha-steroid 4-dehydrogenase 1; 3-oxo-5-alpha-steroid 4-dehydrogenase 1 EC 1.3.99.53-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 1 S5AR 1 SR type 1 Steroid 5-alpha-reductase 15-alpha reductase steroid 5-alpha-reductase type I steroid-5-alpha-reductase alpha polypeptide 1 (3-oxo-5 alpha-steroid delta4-dehydrogenase alpha 1); S5AR 1; SR type 1; Steroid 5-alpha-reductase 1; steroid 5-alpha-reductase type I; steroid-5-alpha-reductase, alpha polypeptide 1 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 1)
Gene Aliases: S5AR 1; SRD5A1
UniProt ID: (Human) P18405
Entrez Gene ID: (Human) 6715
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.