Proteintech
Manufacturer Code:511141AP150UL
Catalog # 50-173-5906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHe42 bHLHe74 F SRC 1 KAT13A NCoA 1 NCOA1 nuclear receptor coactivator 1 Protein Hin 2 RIP160 SRC 1 SRC1 Steroid receptor coactivator 1; bHLHe74; Class E basic helix-loop-helix protein 74; Hin-2 protein; mNRC-1; NCoA-1; Nuclear receptor coactivator 1; Nuclear receptor coactivator protein 1; PAX3/NCOA1 fusion protein; Protein Hin-2; Renal carcinoma antigen NY-REN-52; RIP160; SRC-1; SRC-a/NCoA-1; Steroid receptor coactivator 1; steroid receptor coactivator-1
Gene Aliases: bHLHe42; BHLHE74; F-SRC-1; KAT13A; NCOA1; RIP160; SRC-1; SRC1
UniProt ID: (Human) Q15788, (Mouse) P70365
Entrez Gene ID: (Human) 8648, (Mouse) 17977
Molecular Function: acetyltransferase transcription factor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.