Novus Biologicals
Manufacturer Code:NBP294910100UL
Catalog # NB024326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-198 of human SRI (NP_003121.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 22 kDa protein; calcium binding protein amplified in mutlidrug-resistant cells; calcium binding protein amplified in mutlidrug-resistant cells CP22 CP-22 FLJ2625922 kDa protein H_RG167B05.1 SCN sorcin V19; CP-22; CP22; H_RG167B05.1; Sorcin; V19
Gene Aliases: CP-22; CP22; SCN; SRI; V19
UniProt ID: (Human) P30626
Entrez Gene ID: (Human) 6717
Molecular Function:
annexin
calcium-binding protein
calmodulin
cysteine protease
hydrolase
intracellular calcium-sensing protein
protease
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.