Novus Biologicals
Manufacturer Code:NB400134SS
Catalog # NB400134SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A peptide from the extracellular domain (residues 230-380) of Scavenger Receptor-BI/BII that was expressed as two tandem copies in bacteria using the pET system. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD36; CD36 and LIMPII analogous 1; CD36 and LIMPII analogous 1 CD36 antigen CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 CD36L1scavenger receptor class B type 1 CLA1CD36 antigen-like 1 CLA-1HDLQTL6 Collagen type I receptor thrombospondin receptor-like 1 scavenger receptor class B type III scavenger receptor class B member 1 SRB1scavenger receptor class B member 1 SR-BIMGC138242; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen-like 1; CLA-1; Collagen type I receptor, thrombospondin receptor-like 1; Scavenger receptor class B member 1; scavenger receptor class B type III; scavenger receptor class B, member 1; SR-BI; SRB1
Gene Aliases: CD36L1; CLA-1; CLA1; HDLQTL6; SCARB1; SR-BI; SRB1
UniProt ID: (Human) Q8WTV0
Entrez Gene ID: (Human) 949
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.